Learn More
Invitrogen™ DNA-PK Recombinant Rabbit Monoclonal Antibody (23GB2055), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553211
 View more versions of this product
Product Code. 30089975
  • € 207.00 / 20µL
Description
DNA-PK is a heterotrimeric, DNA-dependent protein kinase that initiates signaling cascades in response to DNA damage. DNA-PK is an atypical kinase, closely related to the PI3 kinases, and is often potently inhibited by small molecules designed to target PI3 kinases. DNA-PK encodes a component of the mediator complex, a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. Multiple transcript variants encoding different isoforms have been found for DNA-PK.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DNA-PK
Recombinant Monoclonal
Unconjugated
PRKDC
AI326420; AU019811; DNA-dependent protein kinase catalytic subunit; DNAPDcs; DNAPK; DNA-PK catalytic subunit; DNA-PKcs; DNPK1; DOXNPH; doxorubicin nephropathy; dxnph; hyper-radiosensitivity of murine scid mutation, complementing 1; HYRC; HYRC1; I79_017613; IMD26; p350; p460; Prkdc; protein kinase, DNA activated, catalytic polypeptide; protein kinase, DNA-activated, catalytic polypeptide; scid; severe combined immunodeficiency; slip; Xrcc7
Rabbit
Affinity chromatography
RUO
5591
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB2055
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P78527
PRKDC
A synthesized peptide derived from human DNA-PKcs (4050-4128AA).