missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA-PKcs, p Ser2056 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05444
This item is not returnable.
View return policy
Description
DNA-PKcs Polyclonal antibody specifically detects DNA-PKcs in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| DNA-PKcs | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| DNA-dependent protein kinase, DNA-dependent protein kinase catalytic subunit-like, DNA-PK catalytic subunit, DNA-PKcs, DNPK1, EC 2.7.11.1, HYRC, HYRC1, p460 | |
| A synthetic phosphorylated peptide around S2056 of human DNA-PKcs (NP_008835.5). SYSSQDPRPA | |
| 100 μg | |
| Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, DNA replication Transcription Translation and Splicing, Non homologous end joining, Phospho Specific, Protein Kinase | |
| 5591 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction