Learn More
DNA Polymerase epsilon subunit 3 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-60438PE
Product Code. 18515722
  • € 615.00 / 0.10mL
Description
DNA Polymerase epsilon subunit 3 Polyclonal antibody specifically detects DNA Polymerase epsilon subunit 3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)