Learn More
Thermo Scientific™ DNA Polymerase I Large (Klenow Fragment) Kit
Product Code. 15496699
Click to view available options
Quantity:
150 units
500 units
Unit Size:
150 units
500 units
Product Code. 15496699
Brand: Thermo Scientific™ J65056.!U
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C
Form Liquid
Format Plastic bottle
Quantity 500 units