missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase Kappa Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-06559AF594
This item is not returnable.
View return policy
Description
DNA Polymerase Kappa Polyclonal antibody specifically detects DNA Polymerase Kappa in Human samples. It is validated for Western Blot
Specifications
| DNA Polymerase Kappa | |
| Polyclonal | |
| Western Blot | |
| DINB protein, DINPPOLQDINB1EC 2.7.7.7, DNA polymerase kappa, polymerase (DNA directed) kappa | |
| The immunogen recognized by this antibody maps to a region between residue 825 and 870 of human DNA polymerase kappa using the numbering given in entry NP_057302.1 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 51426 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction