missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA/RNA Damage Antibody (15A3), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-96878AF488
This item is not returnable.
View return policy
Description
DNA/RNA Damage Monoclonal specifically detects DNA/RNA Damage in Human, Mouse, Non-species specific samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| DNA/RNA Damage | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| 8-hydroxy-guanosine-BSA and-casein conjugates | |
| 0.1 mL | |
| Recognizes markers of oxidative damage to DNA (8-hydroxy-2'-deoxyguanosine, 8-hydroxyguanine and 8-hydroxyguanosine). | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 15A3 | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Human, Mouse, Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction