Learn More
DNA-RNA Hybrid Antibody (S9.6), Novus Biologicals™
Product Code. 30300934 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30300934 100 μg 100µg
30303464 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30300934 Supplier Novus Biologicals Supplier No. NBP344628100ug
Mouse Monoclonal Antibody
DNA-RNA Hybrid Monoclonal antibody specifically detects DNA-RNA Hybrid in All samples. It is validated for ChIP assay,Flow Cytometry,Immunoprecipitation,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Antigen DNA-RNA Hybrid
Applications ChIP Assay, Flow Cytometry, Immunoprecipitation, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone S9.6
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation 10 mM PBS with 0.05% BSA
Host Species Mouse
Immunogen DNA-RNA heteropolymer duplex prepared by transcription of phi X174 single-stranded DNA with DNA-dependent RNA polymerase
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species
Content And Storage Store at 4⪚C.
Form Purified
Isotype IgG2a κ