Learn More
Invitrogen™ DNA/RNA Oxidative Damage Markers Monoclonal Antibody (15A3)
GREENER_CHOICE
Product Code. 11528892
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11528892 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11528892 Supplier Invitrogen™ Supplier No. MA110602
Mouse Monoclonal Antibody
MA1-10602 detects DNA and RNA Oxidative Damage Markers from oxidatively damaged eucaryotic DNA samples. MA1-10602 has been successfully used in ELISA and immunohistochemistry applications. The MA1-10602 immunogen is 8-hydroxyguanosine, conjugated to BSA and casein.
TRUSTED_SUSTAINABILITY
Specifications
Antigen DNA/RNA Oxidative Damage Markers
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 15A3
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.1% sodium azide; pH 7.4
Gene Alias DNA/RNA Damage
Host Species Mouse
Immunogen 8-hydroxyguanosine, conjugated to BSA and casein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Multi-species
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b