missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNA2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA577943

Product Code. 16336654

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Predicted Reactivity: Bovine (84%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

DNA2 is a conserved helicase/nuclease involved in themaintenance of mitochondrial and nuclear DNA stability.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DNA2
Polyclonal
Unconjugated
DNA2
DNA replication ATP-dependent helicase DNA2; DNA replication ATP-dependent helicase/nuclease DNA2; DNA replication ATP-dependent helicase-like homolog; DNA replication helicase 2 homolog; DNA replication helicase 2 homolog (yeast); DNA replication helicase 2-like; DNA replication helicase/nuclease 2; DNA replication nuclease DNA2; DNA2; DNA2L; DNA2-like helicase; E130315B21Rik; hDNA2; Kiaa0083; LOW QUALITY PROTEIN: DNA replication ATP-dependent helicase/nuclease DNA2
Rabbit
Antigen affinity chromatography
RUO
1763
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.75 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
P51530
DNA2
Recombinant protein encompassing a sequence within the N-terminus region of human DNA replication helicase 2. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.