missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557826
This item is not returnable.
View return policy
Description
Immunogen sequence: RIVKVMDDYQ VMDEFLYNLS SDDFNDKWIA SNWPSKILGQ IELVQQQHVE DEEKFRKIQI MDQNNFQEKL EGLQLVVAGF S Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.
This gene encodes an inner dynein arm heavy chain that provides structural support between the radial spokes and the outer doublet of the sperm tail. Naturally occurring mutations in this gene are associated with primary ciliary dyskinesia and multiple morphological anomalies of the flagella that result in asthenozoospermia and male infertility. Mice with a homozygous knockout of the orthologous gene are viable but have reduced sperm motility and are infertile.
Specifications
| DNAH1 | |
| Polyclonal | |
| Unconjugated | |
| DNAH1 | |
| axonemal beta dynein heavy chain 1; Ciliary dynein heavy chain 1; DHC7; Dlp1; DNAH1; DNAH1 variant protein; Dnahc1; dynein axonemal heavy chain 1; dynein heavy chain 1, axonemal; dynein, axonemal, heavy chain 1; dynein, axonemal, heavy polypeptide 1; hDHC7; Heat shock regulated protein 1; HL11; HL-11; HSRF-1; KIAA1410; RGD1311110; testicular tissue protein Li 60; XLHSRF-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25981 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P2D7 | |
| DNAH1 | |
| Recombinant protein corresponding to Human DNAH1. Recombinant protein control fragment (Product #RP-96323). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction