missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNAH10 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA558645

Product Code. 15756521

  • € 594.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SRLTLIEAIN LFKYPAAKSE EELPGVKEFF EHIERERASD VDHMVRWYLA IGPLLTKVEG LVVHTNTGKA PKLASYYKYW EKKIYEVLTK LILKNLQ Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 20%.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

DNAH10
Polyclonal
Unconjugated
DNAH10
Axonemal beta dynein heavy chain 10; ciliary dynein heavy chain 10; DNAH10; Dnahc10; dynein axonemal heavy chain 10; dynein heavy chain 10, axonemal; dynein, axon, heavy chain 10; dynein, axonemal, heavy chain 10; dynein, axonemal, heavy polypeptide 10; KIAA2017
Rabbit
Antigen affinity chromatography
RUO
196385
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8IVF4
DNAH10
Recombinant protein corresponding to Human DNAH10. Recombinant protein control fragment (Product #RP-97884).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.