missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAH6 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30073-1-AP-20UL
This item is not returnable.
View return policy
Description
The DNAH6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human DNAH6. This antibody recognizes human, mouse antigen. The DNAH6 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| DNAH6 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1768 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 500 μg/mL | |
| DKFZp667K087; DNHL1; Dnahc6; FLJ23529; FLJ37357; HL-2; HL2; KIAA1697; MGC132544; MGC141984; MGC177197 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| 475 kDa | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion