missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599618
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Dynein IC1.
Eukaryotic cells rely on actin and microtubule-based protein motors to generate intracellular movements. These protein motors contain specialized domains that hydrolyze ATP to produce force and movement along a cytoskeletal polymer (actin in the case of myosin family and microtubules in the case of the kinesin family and dyneins). Dynein has been implicated in cytoplasmic motile functions, including chromosomal movement, retrograde organelle and axonal transport, the endocytic pathway, and the organization of the Golgi apparatus. In all cell types, dynein has the same basic structures and is composed of two or three distinct heavy chains (~450 kDa), three intermediate chains (70-125 kDa), and at least four light chains (15-25 kDa).
Specifications
| DNAI1 | |
| Polyclonal | |
| Unconjugated | |
| DNAI1 | |
| 1110066F04Rik; axonemal dynein intermediate chain 1; b2b1526Clo; BB124644; CILD1; DIC1; Dnai1; Dnaic1; dynein axonemal intermediate chain 1; Dynein intermediate chain 1, axonemal; dynein, axonemal, intermediate chain 1; dynein, axonemal, intermediate polypeptide 1; I79_025143; ICS; ICS1; immotile cilia syndrome 1; MGC26204; PCD; RP11-296L22.2; testis tissue sperm-binding protein Li 87P | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 27019 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UI46 | |
| DNAI1 | |
| A synthesized peptide derived from human DNAI1(Accession Q9UI46), corresponding to amino acid residues I644-E694. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction