missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJA2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16190
This item is not returnable.
View return policy
Description
DNAJA2 Polyclonal specifically detects DNAJA2 in Human samples. It is validated for Western Blot.
Specifications
| DNAJA2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| cell cycle progression 3 protein, Cell cycle progression restoration gene 3 protein, CPR3PRO3015, Dj3, DJA2, DNAJ, DnaJ (Hsp40) homolog, subfamily A, member 2, dnaJ homolog subfamily A member 2, DNJ3, HIRA interacting protein 4, HIRA-interacting protein 4, HIRIP4RDJ2, Renal carcinoma antigen NY-REN-14 | |
| Rabbit | |
| 46 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 10294 | |
| Human | |
| IgG |
| Western Blot | |
| 0.49mg/mL | |
| Western Blot 1:500-1:3000 | |
| O60884 | |
| DNAJA2 | |
| Recombinant protein encompassing a sequence within the center region of human DNAJA2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Mouse (100%), Chicken (96%), Rat (100%), Xenopus laevis (93%), Bovine (99%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction