missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJA2, Mouse anti-Human, Clone: 2A11-F2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010294-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MANVADTKLYDILGVPPGASENELKKAYRKLAKEYHPDKNPNAGDKFKEISFAYEVLSNPEKRELYDRYGEQGLREGSGGGGGMDDIFSHIFGGGLFGFMGNQSRSRNGRRRGEDMMHPLKVSLEDLYNGKTTKLQLSKNVLCSACSGQGGKSGAVQKCSACRGRGVRIMIRQLAPGMVQQMQSVCSDCNGEGEVINEKDRCKKCEGKKVIKEVKILEVHVDKGMKHGQRITFTGEADQAPGVEPGDIVLLLQEKEHEVFQRDGNDLHMTYKIGLVEALCGFQFTFKHLDGRQIVVKYPPGKVIEPGCVRVVRGEGMPQYRNPFEKGDLYIKFDVQFPENNWINPDRLSELEDLLPSRPEVPNIIGETEEVELQEFDSTRGSGGGQRREAYNDSSDEESSSHHGPGVQCAHQSpecifications
| DNAJA2 | |
| Monoclonal | |
| Unconjugated | |
| DnaJ (Hsp40) homolog, subfamily A, member 2 | |
| CPR3/DJ3/DJA2/DNAJ/DNJ3/HIRIP4/PRO3015/RDJ2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 2A11-F2 | |
| In 1x PBS, pH 7.4 | |
| BC013044 | |
| DNAJA2 | |
| DNAJA2 (AAH13044, 1 a.a. ∼ 412 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10294 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 100UG DNAJA2 monoclonal antibody (M01), clone 2A11 -F2
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium phosphate monobasic, potasssium phosphate dibasic
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction