Learn More
Invitrogen™ DNAJA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596564
Description
Immunogen sequence: FTGEADQAPG VEPGDIVLLL QEKEHEVFQR DGNDLHMTYK IGLVEALCGF QFTFKHLDGR QIVVKYPPGK VIEPGCVRVV RGEGMPQYRN PFEKGDLYIK FDVQFPENNW INPDKLSELE DLLPSRPEVP NIIGETEEVE LQEFDSTRGS GGGQRREAYN DSSDEESSSH HGPGVQCAHQ; Positive Samples: A-375, 293T, HepG2, MCF7, Mouse kidney, Mouse heart, Mouse brain; Cellular Location: Lipid-anchor, Membrane.
The protein encoded by this gene belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain. The product of this gene works as a cochaperone of Hsp70s in protein folding and mitochondrial protein import in vitro.
Specifications
| DNAJA2 | |
| Polyclonal | |
| Unconjugated | |
| DNAJA2 | |
| 1500017M13Rik; 2010206B19Rik; cell cycle progression 3 protein; cell cycle progression restoration gene 3 protein; CPR3; DJ3; DJA2; DNA J protein; DNAJ; DnaJ (Hsp40) homolog, subfamily A, member 2; DnaJ heat shock protein family (Hsp40) member A2; dnaJ homolog subfamily A member 2; Dnaj3; Dnaja2; DNJ3; EGK_12748; HIRA interacting protein 4; HIRA-interacting protein 4; HIRIP4; mDj3; PRO3015; Rdj2; Renal carcinoma antigen NY-REN-14 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10294, 56445, 84026 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 3.79 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35824, O60884, Q9QYJ0 | |
| DNAJA2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 233-412 of human DNAJA2 (NP_0058711). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.