missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565311
This item is not returnable.
View return policy
Description
Immunogen sequence: KGILIIQFLVI FPEKHWLSLE KLPQLEALLP PRQKVRITDD MDQVELKEFC PNEQNWRQHR EAYEEDEDGP QAGVQCQTA Highest antigen sequence identity to the following orthologs - mouse 86%, rat 91%.
DNAJA4 (DnaJ Heat Shock Protein Family (Hsp40) Member A4) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include unfolded protein binding and heat shock protein binding.
Specifications
| DNAJA4 | |
| Polyclonal | |
| Unconjugated | |
| DNAJA4 | |
| 1110021L12Rik; AV358213; Dj4; DnaJ (Hsp40) homolog, subfamily A, member 4; DnaJ heat shock protein family (Hsp40) member A4; dnaJ homolog subfamily A member 4; DNAJA4; heat shock protein, DNAJ-like 4; Hsj4; mmDjA4; MST104; MSTP104; PRO1472 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55466 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WW22 | |
| DNAJA4 | |
| Recombinant Human DNAJA4. Recombinant protein control fragment (Product #a href=. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction