missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 183.00 - € 322.00
Specifications
| Antigen | DNAJB11 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of uLysate, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687698
|
Novus Biologicals
NBP3-12545-50ul |
50 μg |
€ 183.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649996
|
Novus Biologicals
NBP3-12545-100ul |
100 μg |
€ 322.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNAJB11 Polyclonal antibody specifically detects DNAJB11 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| DNAJB11 | |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS, pH7.0 | |
| 51726 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of uLysate, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ABBP-2, APOBEC1-binding protein 2, DnaJ (Hsp40) homolog, subfamily B, member 11, dnaJ homolog subfamily B member 11, DnaJ protein homolog 9, EDJABBP2, ER-associated DNAJ, ER-associated dnaJ protein 3, ER-associated Hsp40 co-chaperone, ERdj3DJ9, ERJ3, ERj3p, hDj9, hDj-9, HEDJERj3, Human DnaJ protein 9, PRO1080, PWP1-interacting protein 4, UNQ537 | |
| Produced in rabbits immunized with E. coli-derived Human DNAJB11 fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title