missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJB12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542571
This item is not returnable.
View return policy
Description
Peptide sequence: ILILILVSAL SQLMVSSPPY SLSPRPSVGH IHRRVTDHLG VVYYVGDTFS Sequence homology: Cow: 93%; Dog: 100%; Goat: 86%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 86%.
DNAJB12 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000).
Specifications
| DNAJB12 | |
| Polyclonal | |
| Unconjugated | |
| DNAJB12 | |
| DJ10; DKFZp586B2023; DnaJ (Hsp40) homolog, subfamily B, member 12; DnaJ heat shock protein family (Hsp40) member B12; DnaJ homolog 10; dnaJ homolog subfamily B member 12; DnaJ subfamily B member 12-like protein; Dnajb12; LOW QUALITY PROTEIN: dnaJ homolog subfamily B member 12; mDj10 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54788 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NXW2 | |
| DNAJB12 | |
| Synthetic peptide directed towards the middle region of human DNAJB12. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction