Learn More
DNAJB2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-32249
Product Code. 18229340
  • ONLINE EXCLUSIVE PRICE
    € 629.00
    € 594.30 / 100µL
    Save € 34.70
    5% Off
Description
DNAJB2 Polyclonal specifically detects DNAJB2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DNAJB2
Polyclonal
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
DnaJ (Hsp40) homolog, subfamily B, member 2, HSJ-1, HSPF3neuronal DNAJ-like 1
Rabbit
Affinity Purified
RUO
3300
Human
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
1.0mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
P25686
DNAJB2
Recombinant protein encompassing a sequence within the center region of human DNAJB2. The exact sequence is proprietary.
100 μL
Primary
Human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%).
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.