missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB2 Antibody (OTI4D12), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70951APC
This item is not returnable.
View return policy
Description
DNAJB2 Monoclonal specifically detects DNAJB2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| DNAJB2 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| DNAJB2 | |
| Full length recombinant protein of human DNAJB2 (NP_001034639) produced in HEK293T cells. | |
| 0.1 mL | |
| 3300.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI4D12 | |
| Flow Cytometry | |
| DnaJ (Hsp40) homolog, subfamily B, member 2, HSJ-1, HSPF3neuronal DNAJ-like 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu