missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB2 Antibody (OTI4D12), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70951DL594
This item is not returnable.
View return policy
Description
DNAJB2 Monoclonal antibody specifically detects DNAJB2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
| DNAJB2 | |
| Monoclonal | |
| DyLight 594 | |
| DnaJ (Hsp40) homolog, subfamily B, member 2, HSJ-1, HSPF3neuronal DNAJ-like 1 | |
| Full length recombinant protein of human DNAJB2 (NP_001034639) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| OTI4D12 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 3300 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction