Learn More
DNAJB2, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand:  Abnova H00003300-D01.100uL
Code produit. 16149664
  • € 365.00 / 100µL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
Sequence: MASYYEILDVPRSASADDIKKAYRRKALQWHPDKNPDNKEFAEKKFKEVAEAYEVLSDKHKREIYDRYGREGLTGTGTGPSRAEAGSGGPGFTFTFRSPEEVFREFFGSGDPFAELFDDLGPFSELQNRGSRHSGPFFTFSSSFPGHSDFSSSSFSFSPGAGAFRSVSTSTTFVQGRRITTRRIMENGQERVEVEEDGQLKSVTINGVPDDLALGLELSRREQQPSVTSRSGGTQVQQTPASCPLDSDLSEDEDLQLAMAYSLSEMEAAGKKPADVF
Spécification
Spécification
DNAJB2
Polyclonal
No additive
NM_001039550.1
DNAJB2
DNAJB2 (NP_001034639.1, 1 a.a. ∼ 277 a.a) full-length human protein.
RUO
3300
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Immunoprecipitation, Western Blot
Unconjugated
DnaJ (Hsp40) homolog, subfamily B, member 2
HSJ1/HSPF3