missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB6 Monoclonal Antibody (OTI3H4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810660S
This item is not returnable.
View return policy
Description
DNAJB6 Monoclonal antibody specifically detects DNAJB6 in Human samples. It is validated for Western BlotSpecifications
| DNAJB6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9HD90 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 58158 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI3H4 | |
| Unconjugated | |
| NEUROD4 | |
| DJ4; DnaJ; DnaJ (Hsp40) homolog, subfamily B, member 6; DnaJ heat shock protein family (Hsp40) member B6; dnaJ homolog subfamily B member 6; DNAJB6; dnajb6 {ECO:0000312; DnaJ-like 2 protein; EMBL:AAH78908.1, ECO:0000312; Heat shock protein J2; HHDJ1; Hsj2; HSJ-2; Hsp40 homolog; LGMD1D; LGMD1E; mammalian relative of DnaJ; mDj4; MRJ; MSJ1; MSJ-1; RGD:1308207} | |
| Full length human recombinant protein of human NEUROD4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction