missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB6, Mouse anti-Human, Clone: 2C11-C1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010049-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MVDYYEVLGVQRHASPEDIKKAYRKLALKWHPDKNPENKEEAERKFKQVAEAYEVLSDAKKRDIYDKYGKEGLNGGGGGGSHFDSPFEFGFTFRNPDDVFREFFGGRDPFSFDFFEDPFEDFFGNRRGPRGSRSRGTGSFFSAFSGFPSFGSGFSSFDTGFTSFGSLGHGGLTSFSSTSFGGSGMGNFKSISTSTKMVNGRKITTKRIVENGQERVEVEEDGQLKSLTINGKEQLLRLDNKSpecifications
| DNAJB6 | |
| Monoclonal | |
| Unconjugated | |
| DnaJ (Hsp40) homolog, subfamily B, member 6 | |
| DJ4/DKFZp566D0824/DnaJ/FLJ42837/HHDJ1/HSJ-2/HSJ2/MGC1152/MGC117297/MRJ/MSJ-1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2C11-C1 | |
| In 1x PBS, pH 7.4 | |
| BC000177 | |
| DNAJB6 | |
| DNAJB6 (AAH00177, 1 a.a. ∼ 241 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10049 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction