missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13157-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene is a member of the J protein family. J proteins function in many cellular processes by regulating the ATPase activity of 70 kDa heat shock proteins. This gene is a member of the type 2 subgroup of DnaJ proteins. The encoded protein is localized to the endoplasmic reticulum. This protein is induced by endoplasmic reticulum stress and plays a role in protecting stressed cells from apoptosis.Specifications
| DNAJB9 | |
| Polyclonal | |
| Unconjugated | |
| DNAJB9 | |
| DNAJB9, ERdj4, Mdg 1, MDG1, MST049, MSTP049 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 24908, 27362, 4189 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P97554, Q9QYI6, Q9UBS3 | |
| DNAJB9 | |
| DNAJB9 Fusion Protein Ag3789 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction