missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNAJC1 Polyclonal Antibody

Product Code. 13452077
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13452077 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13452077 Supplier Invitrogen™ Supplier No. PA527963

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Rhesus Monkey (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The p95 gene (identical to NBS1 and nibrin) is a member of the hMre11/hRad50 double-strand break complex. This protein complex has been implicated in Nijmegen breakage syndrome, an autosomal recessive disorder marked by increased cancer incidence, cell cycle checkpoint deficits, and ionizing radiation sensitivity, thus revealing a direct molecular link between double-strand break repair and cell cycle checkpoint functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene DNAJC1
Gene Accession No. Q96KC8
Gene Alias 4733401K02Rik; AA960110; D230036H06Rik; DnaJ (Hsp40) homolog, subfamily C, member 1; DnaJ (Hsp40) homolog, subfamily C, member 1-like; DnaJ heat shock protein family (Hsp40) member C1; dnaJ homolog subfamily C member 1; DnaJ protein homolog MTJ1; Dnajc1; Dnajl1; DnaJ-like protein; DnaJ-like protein 1; ERdj1; ERj1p; HTJ1; MTJ1
Gene Symbols DNAJC1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DNAJC1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64215
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.