missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC19 Antibody (3H4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00131118-M03
This item is not returnable.
View return policy
Description
DNAJC19 Monoclonal antibody specifically detects DNAJC19 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| DNAJC19 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| DnaJ (Hsp40) homolog, subfamily C, member 19, DnaJ homolog subfamily C member 19, homolog of yeast TIM14, mitochondrial import inner membrane translocase subunit TIM14, Tim14, TIMM14TIM14Pam18 | |
| DNAJC19 (NP_660304, 20 a.a. ∽ 116 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RYVLQAMKHMEPQVKQVFQSLPKSAFSGGYYRGGFEPKMTKREAALILGVSPTANKGKIRDAHRRIMLLNHPDKGGSPYIAAKINEAKDLLEGQAKK | |
| 0.1 mg | |
| Vision | |
| 131118 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 3H4 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| NP_660304 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction