missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJC19 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570454
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
The protein encoded by this gene is thought to be part of a complex involved in the ATP-dependent transport of transit peptide-containing proteins from the inner cell membrane to the mitochondrial matrix. Defects in this gene are a cause of 3-methylglutaconic aciduria type 5 (MGA5), also known as dilated cardiomyopathy with ataxia (DCMA). Alternative splicing of this gene results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 1, 2, 6, 10, 14 and 19. [provided by RefSeq, Jan 2012].
Specifications
| DNAJC19 | |
| Polyclonal | |
| Unconjugated | |
| Dnajc19 | |
| 1810055D05Rik; AA959924; DnaJ (Hsp40) homolog, subfamily C, member 19; DnaJ heat shock protein family (Hsp40) member C19; dnaJ homolog subfamily C member 19; DnaJ homolog, subfamily C, member 19; DNAJC19; DnaJ-like protein subfamily C member 19; homolog of yeast TIM14; hypothetical protein LOC513918; hypothetical protein MGC73251; mitochondrial import inner membrane translocase subunit TIM14; PAM18; RGD1560220; TIM14; TIMM14; zgc:73251 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 67713 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9CQV7 | |
| Dnajc19 | |
| synthetic peptide directed towards the middle region of human DNAJC19 (aa 35-84). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction