missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 204.00 - € 463.00
Specifications
| Antigen | DNAJC2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613992
|
Novus Biologicals
NBP2-92445-0.02ml |
0.02 mL |
€ 204.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18632192
|
Novus Biologicals
NBP2-92445-0.1ml |
0.1 mL |
€ 463.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNAJC2 Polyclonal antibody specifically detects DNAJC2 in Human, Mouse samples. It is validated for Western BlotSpecifications
| DNAJC2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Immunology, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 27000 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DnaJ (Hsp40) homolog, subfamily C, member 2, dnaJ homolog subfamily C member 2, MPHOSPH11M-phase phosphoprotein 11, MPP11Zuotin-related factor 1, ZRF1zuotin, ZUO1, zuotin related factor 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human DNAJC2 (NP_055192.1). MLLLPSAADGRGTAITHALTSASTLCQVEPVGRWFEAFVKRRNRNASASFQELEDKKELSEESEDEELQLEEFPMLKTLDPKDWKNQDHYAVLGLGHVRYKATQRQIKAAHKAMVLKHHPDKRKAAGEPIKEGDNDYFTC | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title