missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC2 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-12803PCP
This item is not returnable.
View return policy
Description
DNAJC2 Polyclonal antibody specifically detects DNAJC2 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| DNAJC2 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| DnaJ (Hsp40) homolog, subfamily C, member 2, dnaJ homolog subfamily C member 2, MPHOSPH11M-phase phosphoprotein 11, MPP11Zuotin-related factor 1, ZRF1zuotin, ZUO1, zuotin related factor 1 | |
| The immunogen this antibody was made to, maps to a region between residue 571 and 621 of human DnaJ (Hsp40) Homolog, Subfamily C, Member 2 using the numbering given in entry NP_055192.1 (GeneID 27000). | |
| 0.1 mL | |
| Cancer, Immunology, Signal Transduction | |
| 27000 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction