missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554212
This item is not returnable.
View return policy
Description
Immunogen sequence: EEINEQIRKE KEEAEARMRQ ASKNTEKSTG GGGNGSKNWS EDDLQLLIKA VNLFPAGTNS RWEVIANYMN IHSSSGVKRT AKDV Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.
EIF4EBP1 is one member of a family of translation repressor proteins. It directly interacts with eukaryotic translation initiation factor 4E (eIF4E), which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5' end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.
Specifications
| DNAJC2 | |
| Polyclonal | |
| Unconjugated | |
| Dnajc2 | |
| (rat homolog) mouse id-associated protein 1; AU020218; DnaJ (Hsp40) homolog, subfamily C, member 2; DnaJ heat shock protein family (Hsp40) member C2; dnaJ homolog subfamily C member 2; DnaJ homolog subfamily C member 2, N-terminally processed; Dnajc2; Gliosarcoma-related antigen MIDA1; Id associate 1; Mida1; Mouse Id associate 1; M-phase phosphoprotein 11; MPHOSPH11; MPP11; Unknown (protein for MGC:139161); ZRF1; Zrf2; ZUO1; zuotin related factor 1; zuotin related factor 2; zuotin-related factor 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116456, 22791, 27000 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P54103, Q7TQ20, Q99543 | |
| Dnajc2 | |
| Recombinant protein corresponding to Human DNAJC2. Recombinant protein control fragment (Product #RP-92635). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction