missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC27 Monoclonal Antibody (OTI8A10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811069S
This item is not returnable.
View return policy
Description
DNAJC27 Monoclonal antibody specifically detects DNAJC27 in Human samples. It is validated for Western BlotSpecifications
| DNAJC27 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y2V3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 30062 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI8A10 | |
| Unconjugated | |
| RAX | |
| AI639580; APAH1; C330021A05Rik; DnaJ (Hsp40) homolog, subfamily C, member 27; DnaJ heat shock protein family (Hsp40) member C27; dnaJ homolog subfamily C member 27; dnajc27; LOW QUALITY PROTEIN: dnaJ homolog subfamily C member 27; rab and DnaJ domain containing; rab and DnaJ domain-containing protein; Rabj; RabJS; Rab-related GTP-binding protein; Ras-associated protein Rap1; RBJ; zgc:194263; zgc:194279 | |
| Human recombinant protein fragment corresponding to amino acids 1-222 of human RAX produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction