missing translation for 'onlineSavingsMsg'
Learn More

DNAJC27 Monoclonal Antibody (OTI9H3), TrueMAB™, OriGene

Product Code. p-200016069
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17564684 30 μL 30µL
17544684 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17564684 Supplier Origene Technologies Supplier No. TA811070S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DNAJC27 Monoclonal antibody specifically detects DNAJC27 in Human samples. It is validated for Western Blot

Specifications

Antigen DNAJC27
Applications Western Blot
Classification Monoclonal
Clone OTI9H3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Dnajc27
Gene Accession No. Q9NZQ0
Gene Alias AI639580; APAH1; C330021A05Rik; DnaJ (Hsp40) homolog, subfamily C, member 27; DnaJ heat shock protein family (Hsp40) member C27; dnaJ homolog subfamily C member 27; dnajc27; LOW QUALITY PROTEIN: dnaJ homolog subfamily C member 27; rab and DnaJ domain containing; rab and DnaJ domain-containing protein; Rabj; RabJS; Rab-related GTP-binding protein; Ras-associated protein Rap1; RBJ; zgc:194263; zgc:194279
Host Species Mouse
Immunogen Full length human recombinant protein of human DNAJC27 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51277
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.