Learn More
DNAJC27 Mouse anti-Human, Clone: OTI7A5, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF811080
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
GTPase which can activate the MEK/ERK pathway and induce cell transformation when overexpressed. May act as a nuclear scaffold for MAPK1, probably by association with MAPK1 nuclear export signal leading to enhanced ERK1/ERK2 signaling.Specifications
| DNAJC27 | |
| Monoclonal | |
| Unconjugated | |
| DNAJC27 | |
| RBJ, RabJS | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51277 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI7A5 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9NZQ0 | |
| DNAJC27 | |
| Full length human recombit protein of human DNAJC27 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.