missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNAJC30 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16307674
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16307674 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16307674 Supplier Invitrogen™ Supplier No. PA580556

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human DNAJC30.

This intronless gene encodes a member of the DNAJ molecular chaperone homology domain-containing protein family. This gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC30
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene DNAJC30
Gene Accession No. Q96LL9
Gene Alias DnaJ (Hsp40) homolog, subfamily C, member 30; DnaJ heat shock protein family (Hsp40) member C30; dnaJ homolog subfamily C member 30; DnaJ homolog subfamily C member 30, mitochondrial; DNAJC30; WBSCR18; Williams Beuren syndrome chromosome region 18; williams-Beuren syndrome chromosomal region 18 protein
Gene Symbols DNAJC30
Host Species Rabbit
Immunogen Recombinant Human DNAJC30 protein (Ser39-Gly124).
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84277
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.