missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC5G Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00285126-B01P
This item is not returnable.
View return policy
Description
DNAJC5G Polyclonal antibody specifically detects DNAJC5G in Human samples. It is validated for Western Blot
Specifications
| DNAJC5G | |
| Polyclonal | |
| Western Blot 1:500 | |
| NP_775921 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 285126 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| CSP-gamma, DnaJ (Hsp40) homolog, subfamily C, member 5 gamma, dnaJ homolog subfamily C member 5G, FLJ40417, gamma cysteine string protein, gamma-CSP, Gamma-cysteine string protein | |
| DNAJC5G (NP_775921, 1 a.a. - 189 a.a.) full-length human protein. MSTVKEAAHRLSKSEMSLYAVLDLKKGASPEDFKKSYSHSALLPHPPFEYHLGRKLALRYHPDKNPGNAQAAEIFKEINAAHAILSDSKKRKIYDQHGSLGIYLYDHFGEEGVRYYFILNSCWFKTLVILCTLLTCCCFCCCCCFCCGALKPPPEQDSGRKYQQNVQSQPPRSGAKCDFRSEENSEDDF | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction