missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNAJC6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13201219
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13201219 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13201219 Supplier Invitrogen™ Supplier No. PA526981

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DNAJC6 Polyclonal Antibody for Flow, IHC (P), Western Blot

DNAJC6 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus, a glycine/phenylalanine (G/F)-rich region, and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC6
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene DNAJC6
Gene Accession No. O75061, Q80TZ3
Gene Alias 2810027M23Rik; auxilin; DJC6; DnaJ (Hsp40) homolog, subfamily B, member 6; DnaJ (Hsp40) homolog, subfamily C, member 6; DnaJ heat shock protein family (Hsp40) member C6; dnaJ homolog subfamily C member 6; DNAJC6; Kiaa0473; mKIAA0473; PARK19; putative tyrosine-protein phosphatase auxilin
Gene Symbols DNAJC6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 254-281 amino acids from the central region of human DNAJC6.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72685, 9829
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.