Learn More
Invitrogen™ DNAJC8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109647
Description
Immunogen sequence: MAASGESGTS GGGGSTEEAF MTFYSEVKQI EKRDSVLTSK NQIERLTRPG SSYFNLNPFE VLQIDPEVTD EEIKKRFRQL SILVHPDKNQ DDADRAQKAF EAVDKAYKLL LDQEQKKRAL DVIQAGKEYV EHTVKERKKQ LKKEGKPTIV EEDDPELFKQ AVYKQTMKLF AELEIKRKER EAKEMHERKR QREEEIEAQE KAKREREWQK NFEESRDGRV DSWRNFQANT KGKKEKKNRT FLRPPKVKME QRE.
Suppresses polyglutamine (polyQ) aggregation of ATXN3 in neuronal cells (PubMed:27133716). [UniProt].
Specifications
| DNAJC8 | |
| Polyclonal | |
| Unconjugated | |
| DNAJC8 | |
| 1110021D09Rik; 2010009J04Rik; AL024084; AU019262; AU044514; DnaJ (Hsp40) homolog, subfamily C, member 8; DnaJ heat shock protein family (Hsp40) member C8; dnaJ homolog subfamily C member 8; DNAJC8; HSPC315; HSPC331; SPF31; splicing protein spf31 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 68598 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.78 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6NZB0 | |
| DNAJC8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-253 of human DNAJC8 (NP_055095.2). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.