missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAM-1/CD226 Antibody (2770B), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB10988Y
This item is not returnable.
View return policy
Description
DNAM-1/CD226 Monoclonal antibody specifically detects DNAM-1/CD226 in Human samples. It is validated for Flow Cytometry
Specifications
| DNAM-1/CD226 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 2770B | |
| Flow Cytometry | |
| CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1 | |
| Human embryonic kidney cell HEK293-derived human DNAM-1/CD226 protein, Glu19-Asn247, Accession # Q15762 | |
| 0.1 mL | |
| Cellular Markers, Innate Immunity, Signal Transduction | |
| 10666 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction