missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAM-1/CD226 Antibody (838216), Alexa Fluor™ 532, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB4436X
This item is not returnable.
View return policy
Description
DNAM-1/CD226 Monoclonal antibody specifically detects DNAM-1/CD226 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| DNAM-1/CD226 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 838216 | |
| Flow Cytometry, CyTOF-ready | |
| CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse DNAM-1/CD226, Met1-Pro254, Accession # Q8K4F0 | |
| 0.1 mL | |
| Cellular Markers, Innate Immunity, Signal Transduction | |
| 10666 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction