missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAM-1/CD226 Antibody (BLR063G), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-77357
This item is not returnable.
View return policy
Description
DNAM-1/CD226 Monoclonal antibody specifically detects DNAM-1/CD226 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| DNAM-1/CD226 | |
| Monoclonal | |
| Unconjugated | |
| Borate Buffered Saline (BBS) pH 8.2 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| BLR063G | |
| Western Blot 1:1000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 20 μg/mg lysate, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
| CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1 | |
| Immunogen was a recombinant protein representing the extracellular domain of human DNAM1/CD226. | |
| 100 μg | |
| Cellular Markers, Innate Immunity, Signal Transduction | |
| 10666 | |
| Store at 2°C to 8°C, 1 year from date of receipt | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction