missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNase II Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97515-100ul
This item is not returnable.
View return policy
Description
DNase II Polyclonal antibody specifically detects DNase II in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| DNase II | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| Acid DNase, Deoxyribonuclease II alpha, deoxyribonuclease II, lysosomal, deoxyribonuclease-2-alpha, DNase II alpha, DNase II, lysosomal, DNASE2A, DNL, DNL2, EC 3.1.22.1, Lysosomal DNase II, R31240_2 | |
| Produced in rabbits immunized with E. coli-derived Human DNase II fragment. | |
| 100 μg | |
| Apoptosis, Cell Biology, Cell Cycle and Replication | |
| 1777 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction