missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNase II Polyclonal Antibody
GREENER_CHOICE

Product Code. 11561503
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11561503 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11561503 Supplier Invitrogen™ Supplier No. PA130778

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: Human spleen tissue lysate. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

DNase II is a member of the DNase family. The protein, located in the lysosome, hydrolyzes DNA under acidic conditions and mediates the breakdown of DNA during erythropoiesis and apoptosis. Two codominant alleles have been characterized, DNASE2*L (low activity) and DNASE2*H (high activity), that differ at one nucleotide in the promoter region. The DNASE2*H allele is represented in this record.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNase II
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Gene DNASE2
Gene Accession No. O00115
Gene Alias acid DNase; deoxyribonuclease; Deoxyribonuclease II alpha; deoxyribonuclease II, lysosomal; deoxyribonuclease-2-alpha; DNase II alpha; DNase II, lysosomal; DNASE2; Dnase2a; DNL; Dnl2; lysosomal DNase II; R31240_2
Gene Symbols DNASE2
Host Species Rabbit
Immunogen Peptide corresponding to aa 347- 360 of human DNase II precursor (accession no. AF047016).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1777
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.