missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNASE2B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-62278
This item is not returnable.
View return policy
Description
DNASE2B Polyclonal antibody specifically detects DNASE2B in Human samples. It is validated for Western Blot
Specifications
| DNASE2B | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| deoxyribonuclease II betaDNase2-like acid DNase, DLADdeoxyribonuclease-2-beta, DNase II beta, DNase II-like acid DNase, EC 3.1.22.1, Endonuclease DLAD, lysosomal DNase II | |
| Synthetic peptides corresponding to DNASE2B(deoxyribonuclease II beta) The peptide sequence was selected form the N terminal of DNASE2B. Peptide sequence EGKAVDWFTFYKLPKRQNKESGETGLEYLYLDSTTRSWRKSEQLMNDTKS. The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μg | |
| Vision | |
| 58511 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1.0 μg/mL | |
| Q8WZ79 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction