missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNCIC1 Antibody (74.1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-726
This item is not returnable.
View return policy
Description
DNCIC1 Monoclonal specifically detects DNCIC1 in Human, Mouse, Rat, Drosophila, Mammal, Xenopus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| DNCIC1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 ug/ml, Flow Cytometry 1 - 2 ug, Immunocytochemistry/Immunofluorescence 2 - 4 ug/ml, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| cytoplasmic dynein 1 intermediate chain 1, Cytoplasmic dynein intermediate chain 1, DNCI1cytoplasmic, intermediate polypeptide 1, DNCIC1DH IC-1, Dynein intermediate chain 1, cytosolic, dynein, cytoplasmic 1, intermediate chain 1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 1780 | |
| Human, Mouse, Rat, Drosophila, Mammalia, Xenopus species | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 74.1 | |
| Unconjugated | |
| O14576 | |
| DYNC1I1 | |
| Purified bovine brain cytoplasmic dynein. | |
| 100 μg | |
| Primary | |
| Detects two isoforms of the 74kDa polypeptide of cytoplasmic Dynein. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction