missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNCIC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38053-20ul
This item is not returnable.
View return policy
Description
DNCIC1 Polyclonal antibody specifically detects DNCIC1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| DNCIC1 | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:200 | |
| cytoplasmic dynein 1 intermediate chain 1, Cytoplasmic dynein intermediate chain 1, DNCI1cytoplasmic, intermediate polypeptide 1, DNCIC1DH IC-1, Dynein intermediate chain 1, cytosolic, dynein, cytoplasmic 1, intermediate chain 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human DNCIC1 (NP_001129028.1).,, Sequence:, MSDKSDLKAELERKKQRLAQIREEKKRKEEERKKKEADMQQKKEPVQDDSDLDRKRRETEALLQSIGISPEPPLVPTPMSPSSKSVSTPSEAGSQDSGDLGPLTRTLQWDTDPSVLQLQSDSELGRRLHKLGVSKVTQVDFLPREVVSYSKETQTPLATHQSEEDEEDEEMVESKVGQDSELENQDKKQEVKEAPPRELTEEEKQQILHSEEFLIFFDRT | |
| 20 μL | |
| Cell Biology, Cellular Markers, Golgi Apparatus Markers, Neuronal Cell Markers | |
| 1780 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur