Learn More
Invitrogen™ DNER Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120373
Description
Positive test controls include: Hela, HT-29, Mouse brain, Mouse eye, Mouse small intestine, Rat brain. The target is usually found in the following locations: Cell membrane, Single-pass type I membrane protein. Immunogen sequence: PATGWTESMA PRQLQPVPAT QEPDKILPRS QATVTLPTWQ PKTGQKVVEM KWDQVEVIPD IACGNASSNS SAGGRLVSFE VPQNTSVKIR QDATASLILL W.
DNER (Delta-notch-like EGF-related receptor), also known as Delta-notch-like EGF repeat-containing transmembrane protein, is a neuron-specific, atypical Notch ligand expressed in dendrites and cell bodies of neurons throughout the central nervous system. DNER contains ten extracellular EGF-like domains that are highly homologous to those of the Notch ligand, Delta. In the cerebellum, DNER is predominantly expressed in Purkinje cells. DNER mediates neuron-glia interaction during astrocytogenesis through a direct interaction with Notch 1 at Purkinje cell/Bergmann glia contacts. This interaction activates a Deltex-dependent Notch signaling pathway in Bergmann glia and may regulate Bergmann glial morphogenesis. DNER is crucial for the functional and morphological maturation of Bergmann glia. DNER-knockout mice are characterized by motor discoordination and cerebellum retardation in morphogenesis.
Specifications
| DNER | |
| Polyclonal | |
| Unconjugated | |
| Dner | |
| A930026D19Rik; BET; Brain EGF repeat-containing transmembrane protein; Bret; Delta and Notch-like epidermal growth factor-related receptor; delta/notch like EGF repeat containing; delta/notch-like EGF repeat containing; delta/notch-like EGF-related receptor; delta-notch-like EGF repeat-containing transmembrane; Dner; H_NH0150O02.1; LOW QUALITY PROTEIN: delta and Notch-like epidermal growth factor-related receptor; transmembrane protein Bet; UNQ26; UNQ262/PRO299; WUGSC:H_NH0150O02.1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 227325, 316573, 92737 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.56 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8JZM4, Q8NFT8 | |
| Dner | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 140-240 of human DNER (NP_620711.3). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.