missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MACF1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36528JF646
This item is not returnable.
View return policy
Description
MACF1 Polyclonal antibody specifically detects MACF1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| MACF1 | |
| Polyclonal | |
| Western Blot | |
| microtubule-actin crosslinking factor 1 | |
| The immunogen recognized by this antibody maps to a region between residue 1975 to 2025 of human Microtubule-Actin Crosslinking Factor 1 using the numbering given in entry NP_036222.3. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 23499 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction