Learn More
DNMT1 Antibody (4G11-C7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59344
 View more versions of this product
Product Code. 18217211
  • € 559.00 / 100µg
Description
DNMT1 Monoclonal specifically detects DNMT1 in Human, Mouse, Fish, Zebrafish samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DNMT1
Monoclonal
Unconjugated
PBS pH7.4, 50% glycerol with 0.09% Sodium Azide
DNMT1
Raised against a partial protein corresponding to amino acids 620-950 of human DNMT1
100 μg
Angiogenesis, Cancer, Chromatin Modifiers, Chromatin Research, Core ESC Like Genes, DNA Methyltransferases, Epigenetics, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators
1786
Human, Mouse, Fish, Zebrafish
Purified
Western Blot, Immunohistochemistry, Immunoprecipitation, ChIP Assay
4G11-C7
Western Blot 1:1000, Immunohistochemistry 1:1000, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992
Mouse
Protein G purified
RUO
Primary
Detects 180kDa. It will cross-react with mouse DNMT1.
Store at -20C.